Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291041.1 | NP_001277970.1 | 2 | 231 | ||
NM_021782.3 | NP_068554.1 | 2 | 231 | |||
GenBank mRNA | AF254070.1 | - | 2 | 213 | ||
AK155984.1 | - | 1 | 348 | |||
AY428162.1 | - | 2 | 160 | |||
BC125414.1 | - | 2 | 201 | 40129929 | ||
BC125416.1 | - | 2 | 201 | 40129931 | ||
DQ645418.1 | - | 2 | 160 |