Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yeats4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026570.4 | NP_080846.1 | 4, 5 | 528 | ||
GenBank mRNA | AK002839.1 | - | 3, 4 | 176 | ||
AK010522.1 | - | 4, 5 | 529 | |||
AK012096.1 | - | 4, 5 | 495 | |||
AK081867.1 | - | 4, 5 | 539 | |||
AK219066.1 | - | 3, 4 | 181 | |||
BC020043.1 | - | 4, 5 | 496 | 4013548 |