Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290437.1 | NP_001277366.1 | 9, 10 | 2022 | ||
NM_033134.3 | NP_149125.1 | 8, 9 | 2306 | |||
GenBank mRNA | AF226683.1 | - | 8, 9 | 2268 | ||
AK047245.1 | - | 6, 7 | 2235 | |||
AK080075.1 | - | 8, 9 | 2288 | |||
AK085439.1 | - | 9, 10 | 2027 | |||
AK154097.1 | - | 8, 9 | 2307 | |||
AK158135.1 | - | 8, 9 | 2291 | |||
AK170174.1 | - | 8, 9 | 2376 | |||
AK170786.1 | - | 8, 9 | 2263 | |||
AK170812.1 | - | 7, 8 | 2341 | |||
BC052717.1 | - | 8, 9 | 2275 | 6837339 | ||
BC057566.1 | - | 8, 9 | 2358 | |||
BC080295.1 | - | 8, 9 | 2271 | 30354397 |