Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033134.3 | NP_149125.1 | 10 | 2467 | ||
GenBank mRNA | AF226683.1 | - | 10 | 2429 | ||
AK047245.1 | - | 8 | 2396 | |||
AK080075.1 | - | 10 | 2449 | |||
AK154097.1 | - | 10 | 2468 | |||
AK158135.1 | - | 10 | 2452 | |||
AK170174.1 | - | 10 | 2537 | |||
AK170786.1 | - | 10 | 2424 | |||
AK170812.1 | - | 9 | 2502 | |||
BC052717.1 | - | 10 | 2436 | 6837339 | ||
BC057566.1 | - | 10 | 2519 | |||
BC080295.1 | - | 10 | 2432 | 30354397 |