Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023219.2 | NP_075708.2 | 5 | 499 | ||
GenBank mRNA | AF251268.1 | - | 5 | 499 | ||
AF411960.2 | - | 5 | 419 | |||
AK144296.1 | - | 442 | ||||
BC109133.1 | - | 5 | 438 | |||
BC109134.1 | - | 5 | 438 | |||
BC127612.1 | - | 5 | 438 | |||
BC127615.1 | - | 5 | 438 | 40061657 | ||
BC128465.1 | - | 5 | 438 |