Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022657.4 | NP_073148.1 | 3 | 630 | ||
GenBank mRNA | AF263536.1 | - | 3 | 558 | ||
AK155687.1 | - | 3 | 630 |