Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168240.1 | NP_001161712.1 | 3 | 800 | ||
NM_022986.4 | NP_075362.3 | 4 | 866 | |||
GenBank mRNA | AF093135.1 | - | 4 | 649 | ||
AJ242722.1 | - | 4 | 491 | |||
AK009948.1 | - | 2 | 241 | |||
AK013036.2 | - | 4 | 671 | |||
AK014712.1 | - | 3 | 577 | |||
AK020448.1 | - | 4 | 641 | |||
AK045436.1 | - | 4 | 662 | |||
AK076594.1 | - | 4 | 674 | |||
BC046301.1 | - | 4 | 660 | |||
BC061099.1 | - | 4 | 653 | 6742251 |