Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168240.1 | NP_001161712.1 | 3 | 877 | ||
NM_022986.4 | NP_075362.3 | 4 | 943 | |||
GenBank mRNA | AF093135.1 | - | 4 | 726 | ||
AK009948.1 | - | 2 | 318 | |||
AK013036.2 | - | 4 | 748 | |||
AK014712.1 | - | 3 | 654 | |||
AK020448.1 | - | 4 | 718 | |||
AK045436.1 | - | 4 | 739 | |||
AK076594.1 | - | 4 | 751 | |||
BC046301.1 | - | 4 | 737 | |||
BC061099.1 | - | 4 | 730 | 6742251 |