Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164533.1 | NP_001158005.1 | 5 | 770 | ||
NM_022994.3 | NP_075370.2 | 5 | 770 | |||
GenBank mRNA | AJ250375.1 | - | 5 | 514 | ||
AK014280.1 | - | 5 | 440 | |||
AK014889.1 | - | 5 | 440 | |||
AK032275.1 | - | 5 | 442 | |||
AK088960.1 | - | 5 | 440 | |||
BC019566.1 | - | 5 | 770 | 4235517 |