Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbata | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017407.1 | NP_001017407.1 | 4 | 589 | ||
NM_001017409.1 | NP_001017409.1 | 4 | 656 | |||
NM_001017419.2 | NP_001017419.1 | 5 | 671 | |||
NM_001017433.2 | NP_001017433.1 | 5 | 773 | |||
NM_001017441.2 | NP_001017441.2 | 5 | 671 | |||
NM_023064.3 | NP_075551.3 | 4 | 691 | |||
GenBank mRNA | AF257502.1 | - | 4 | 691 | ||
AF257503.1 | - | 4 | 589 | |||
AF521591.1 | - | 4 | 602 | |||
AF521592.1 | - | 4 | 704 | |||
AJ249392.1 | - | 3 | 628 | |||
AK006213.1 | - | 3 | 236 | |||
AK038284.1 | - | 3 | 459 | |||
AK088524.1 | - | 3 | 560 | |||
AK132793.1 | - | 5 | 672 | |||
AY243457.1 | - | 4 | 658 | |||
AY243458.1 | - | 3 | 438 | |||
AY243459.1 | - | 3 | 627 | |||
EU622858.1 | - | 4 | 555 | |||
X83884.1 | - | 1 | 40 |