Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025334.3 | NP_079610.1 | 6 | 495 | ||
GenBank mRNA | AK002862.1 | - | 6 | 482 | ||
AK003481.1 | - | 6 | 476 | |||
AY548113.1 | - | 6 | 407 | |||
BC006857.1 | - | 6 | 495 | 2615934 |