Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025341.3 | NP_079617.2 | 2 | 109 | ||
GenBank mRNA | AK002883.1 | - | 2 | 69 | ||
AK076105.1 | - | 2 | 188 | |||
AK090076.1 | - | 2 | 109 | |||
BC027011.1 | - | 2 | 102 | 5369937 | ||
BC051086.1 | - | 2877 |