Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025383.3 | NP_079659.1 | 3 | 317 | ||
GenBank mRNA | AK003463.1 | - | 3 | 284 | ||
AK145874.1 | - | 3 | 305 | |||
AK155871.1 | - | 3 | 305 | |||
AK156566.1 | - | 3 | 294 | |||
AK170987.1 | - | 3 | 302 | |||
AK172538.1 | - | 3 | 287 | |||
AK213029.1 | - | 3 | 209 | |||
AK218572.1 | - | 3 | 226 | |||
BC037069.1 | - | 3 | 316 | 5002765 |