Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023168.3 | NP_075657.1 | 5 | 747 | ||
GenBank mRNA | AF182040.1 | - | 5 | 730 | ||
AK187074.1 | - | 2 | 129 | |||
AK188174.1 | - | 2 | 48 | |||
AK189399.1 | - | 4 | 371 | |||
AK218437.1 | - | 2 | 48 | |||
BC010802.1 | - | 5 | 780 | 4236429 | ||
BC019157.1 | - | 5 | 768 | |||
BC037667.1 | - | 5 | 747 | 5376054 |