Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025408.2 | NP_079684.2 | 11 | 855 | ||
GenBank mRNA | AK004287.1 | - | 11 | 856 | ||
AK011668.1 | - | 11 | 836 | |||
AK080977.1 | - | 11 | 833 | |||
AK150629.1 | - | 11 | 816 | |||
AK162403.1 | - | 11 | 871 | |||
BC023924.1 | - | 12 | 1023 | 5324078 |