Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025408.2 | NP_079684.2 | 7, 8 | 584 | ||
GenBank mRNA | AK004287.1 | - | 7, 8 | 585 | ||
AK011668.1 | - | 7, 8 | 565 | |||
AK017361.1 | - | 489 | ||||
AK080977.1 | - | 7, 8 | 562 | |||
AK150629.1 | - | 7, 8 | 545 | |||
AK162403.1 | - | 7, 8 | 600 | |||
AK171957.1 | - | 1 | 398 | |||
BC023924.1 | - | 8, 9 | 752 | 5324078 |