Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291197.1 | NP_001278126.1 | 6 | 676 | ||
NM_001291201.1 | NP_001278130.1 | 5 | 616 | |||
NM_001291202.1 | NP_001278131.1 | 5 | 613 | |||
NM_025482.3 | NP_079758.2 | 6 | 673 | |||
GenBank mRNA | AK013085.1 | - | 6 | 615 | ||
AK013188.1 | - | 6 | 626 | |||
AK051268.1 | - | 5 | 557 | |||
AK159925.1 | - | 6 | 601 | |||
BC006886.1 | - | 6 | 562 | 2647702 | ||
DV047279.1 | - | 6 | 662 |