Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025375.2 | NP_001020546.1 | 11 | 1124 | ||
NM_001025376.2 | NP_001020547.1 | 11 | 1013 | |||
NM_023191.3 | NP_075680.1 | 11 | 1018 | |||
GenBank mRNA | AF309554.1 | - | 11 | 976 | ||
AK010283.1 | - | 11 | 964 | |||
AK012342.1 | - | 11 | 1030 | |||
AK013114.1 | - | 11 | 982 | |||
AK142498.1 | - | 11 | 981 | |||
AK153862.1 | - | 11 | 1051 | |||
AK167706.1 | - | 11 | 1051 | |||
AK219807.1 | - | 3 | 179 | |||
BC023026.1 | - | 11 | 963 | 5374890 | ||
EF011623.1 | - | 11 | 961 |