Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025495.3 | NP_079771.1 | 1 | 107 | ||
GenBank mRNA | AK006234.1 | - | 1 | 145 | ||
AK012863.1 | - | 1 | 111 | |||
AK014919.1 | - | 1 | 63 | |||
AK076577.1 | - | 1 | 145 | |||
BC094280.1 | - | 1 | 161 | |||
BC147207.1 | - | 1 | 87 | 9056262 | ||
BC147208.1 | - | 1 | 87 | 9056260 |