Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025495.3 | NP_079771.1 | 5, 6 | 604 | ||
GenBank mRNA | AK006234.1 | - | 5, 6 | 641 | ||
AK012863.1 | - | 5, 6 | 608 | |||
BC094280.1 | - | 5, 6 | 657 | |||
BC147207.1 | - | 5, 6 | 584 | 9056262 | ||
BC147208.1 | - | 5, 6 | 584 | 9056260 |