Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025535.2 | NP_079811.1 | 3 | 246 | ||
GenBank mRNA | AK002327.1 | - | 3 | 223 | ||
AK010187.1 | - | 3 | 229 | |||
AK013559.1 | - | 3 | 208 | |||
AK150906.1 | - | 3 | 202 | |||
AK155429.1 | - | 3 | 216 | |||
BC082550.1 | - | 3 | 215 | 30357800 | ||
BC085178.1 | - | 3 | 199 | 30701201 | ||
BY068863.1 | - | 3 | 246 |