Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170592.2 | NP_733480.1 | 2 | 171 | ||
GenBank mRNA | AK009056.1 | - | 2 | 181 | ||
AK011888.1 | - | 2 | 181 | |||
AK011960.1 | - | 2 | 173 | |||
AK154783.1 | - | 2 | 181 | |||
AK184965.1 | - | 1 | 26 | |||
AK218665.1 | - | 1 | 26 | |||
BC027220.1 | - | 2 | 171 | 3991341 |