Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025673.2 | NP_079949.1 | 4 | 1073 | ||
GenBank mRNA | AB271918.1 | - | 2 | 826 | ||
AK005996.1 | - | 4 | 1120 | |||
AK015543.1 | - | 4 | 1117 | |||
AK028409.1 | - | 4 | 1084 | |||
AK077446.1 | - | 4 | 1138 | |||
AK133763.1 | - | 4 | 1121 | |||
AK136194.1 | - | 4 | 698 | |||
AK139545.1 | - | 4 | 1120 | |||
BC003961.1 | - | 1 | 164 | |||
BC027333.1 | - | 1 | 1269 | |||
BC031445.1 | - | 5 | 983 | 4482612 | ||
BC039909.1 | - | 4 | 1070 | |||
BC051035.1 | - | 4 | 1073 | 2655316 | ||
BC058233.1 | - | 1 | 32 |