Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025724.2 | NP_080000.1 | 1 | 892 | ||
GenBank mRNA | AK006212.1 | - | 1 | 893 | ||
AK014860.1 | - | 1 | 893 | |||
BC120881.1 | - | 1 | 817 | 40057872 | ||
BC120882.1 | - | 1 | 817 | 40057873 |