Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdelr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025841.4 | NP_080117.1 | 3 | 402 | ||
GenBank mRNA | AJ278133.1 | - | 3 | 395 | ||
AK003515.1 | - | 3 | 397 | |||
AK007403.1 | - | 3 | 386 | |||
AK150402.1 | - | 3 | 397 | |||
BC007146.1 | - | 3 | 384 | 3496891 | ||
CT010232.1 | - | 3 | 260 |