Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025846.2 | NP_080122.2 | 5 | 787 | ||
GenBank mRNA | AK003733.1 | - | 2 | 173 | ||
AK011419.1 | - | 5 | 787 | |||
AK078870.1 | - | 5 | 788 | |||
AK172037.1 | - | 5 | 779 | |||
AK172113.1 | - | 5 | 2059 | |||
BC003871.1 | - | 5 | 738 | 3492159 |