Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175113.3 | NP_780322.2 | 7 | 1006 | ||
GenBank mRNA | AK014359.1 | - | 7 | 1122 | ||
AK017672.1 | - | 7 | 948 | |||
AK028755.1 | - | 7 | 1006 | |||
AK129300.1 | - | 7 | 973 | |||
AK164396.1 | - | 7 | 939 | |||
AK165976.1 | - | 7 | 958 | |||
AK167222.1 | - | 7 | 972 | |||
BC049083.1 | - | 4 | 763 | |||
BC052648.1 | - | 7 | 924 | 30030801 |