Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025862.2 | NP_080138.2 | 3, 4 | 418 | ||
GenBank mRNA | AK009379.1 | - | 3, 4 | 397 | ||
AK019502.1 | - | 3, 4 | 415 | |||
AK029897.1 | - | 3, 4 | 382 | |||
AK051091.1 | - | 3, 4 | 373 | |||
AK077335.1 | - | 3, 4 | 398 | |||
AK161255.1 | - | 3, 4 | 372 | |||
AK211554.1 | - | 1, 2 | 137 | |||
BC026535.1 | - | 3, 4 | 392 | |||
BC037644.1 | - | 3, 4 | 394 | 5053565 | ||
BQ931973.1 | - | 3, 4 | 418 |