Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025891.3 | NP_080167.3 | 8 | 1018 | ||
GenBank mRNA | AK005094.1 | - | 8 | 965 | ||
AK019374.1 | - | 1 | 99 | |||
AK158019.1 | - | 988 | ||||
BC013122.1 | - | 8 | 1055 | 3586276 | ||
BC060525.1 | - | 8 | 917 | 6817219 |