Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025911.2 | NP_080187.2 | 11 | 1153 | ||
GenBank mRNA | AK007917.1 | - | 11 | 1154 | ||
BC006734.1 | - | 11 | 1058 | 3156328 | ||
BC024946.1 | - | 6 | 411 | 4010938 | ||
BC049073.1 | - | 11 | 1122 | 5707404 |