Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025928.3 | NP_080204.1 | 3 | 322 | ||
GenBank mRNA | AF348510.1 | - | 3 | 332 | ||
AK011174.1 | - | 3 | 324 | |||
AK012113.1 | - | 3 | 323 | |||
AK013787.1 | - | 3 | 325 | |||
AK078308.1 | - | 3 | 341 | |||
AK168200.1 | - | 3 | 323 | |||
AK180171.1 | - | 1 | 57 | |||
BC034333.1 | - | 3 | 300 | |||
BC037139.1 | - | 3 | 331 | 4952550 | ||
BC049223.1 | - | 3 | 321 | 3257320 |