Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025964.3 | NP_080240.1 | 3 | 1320 | ||
GenBank mRNA | AK009673.1 | - | 4 | 640 | ||
AK009675.1 | - | 4 | 642 | |||
AK009678.1 | - | 4 | 642 | |||
AK009679.1 | - | 4 | 642 | |||
AK009680.1 | - | 4 | 642 | |||
AK009682.1 | - | 4 | 642 | |||
AK011433.1 | - | 4 | 666 | |||
AK033578.1 | - | 3 | 1321 | |||
BC026952.1 | - | 3 | 590 | 4483032 |