Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026012.2 | NP_080288.1 | 3, 4 | 419 | ||
GenBank mRNA | AF534394.1 | - | 2, 3 | 231 | ||
AK007483.1 | - | 1, 2 | 76 | |||
AK160222.1 | - | 3, 4 | 405 | |||
BC039808.1 | - | 3, 4 | 389 | 1547029 | ||
BC069856.1 | - | 3, 4 | 419 | 6491606 |