Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026033.2 | NP_080309.2 | 4 | 905 | ||
GenBank mRNA | AB047921.1 | - | 4 | 886 | ||
AK078416.1 | - | 4 | 850 | |||
BC019449.1 | - | 4 | 775 | 4024452 |