Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024196.3 | NP_077158.1 | 6 | 807 | ||
GenBank mRNA | AK006877.1 | - | 6 | 804 | ||
AK036751.1 | - | 719 | ||||
AK076124.1 | - | 6 | 808 | |||
AK082369.1 | - | 957 | ||||
AK085702.1 | - | 805 | ||||
AK158274.1 | - | 804 | ||||
AK168985.1 | - | 808 | ||||
AK172349.1 | - | 804 | ||||
AK172372.1 | - | 236 | ||||
BC002196.1 | - | 5 | 623 | |||
BC034504.1 | - | 789 | 4192736 | |||
BC040089.1 | - | 6 | 770 | 3991310 | ||
BC086617.1 | - | 793 |