Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026212.2 | NP_080488.1 | 2, 3 | 531 | ||
GenBank mRNA | AK010891.1 | - | 2, 3 | 340 | ||
AK090271.1 | - | 2, 3 | 533 | |||
AY072769.1 | - | 2, 3 | 324 | |||
BC125530.1 | - | 2, 3 | 405 | 40130045 | ||
BC137840.1 | - | 2, 3 | 405 | 8860859 |