Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026329.2 | NP_080605.1 | 3 | 327 | ||
GenBank mRNA | AK007892.1 | - | 3 | 349 | ||
AK010687.1 | - | 3 | 296 | |||
AK011627.1 | - | 3 | 328 | |||
AK013134.1 | - | 3 | 298 | |||
AK017473.1 | - | 3 | 306 | |||
BC005580.1 | - | 3 | 346 | 3968561 | ||
BC055278.1 | - | 3 | 277 | 5699685 |