Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026341.2 | NP_080617.2 | 7 | 722 | ||
GenBank mRNA | AK014204.1 | - | 8 | 945 | ||
AK017031.1 | - | 7 | 691 | |||
BC037091.1 | - | 6 | 645 | |||
BC040807.1 | - | 7 | 669 | |||
CA983915.1 | - | 5 | 458 | |||
CX219422.1 | - | 2 | 130 |