Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026341.2 | NP_080617.2 | 8 | 925 | ||
GenBank mRNA | AK014204.1 | - | 9 | 1148 | ||
AK017031.1 | - | 8 | 885 | |||
AK161922.1 | - | 7 | 875 | |||
BC021366.1 | - | 9 | 1079 | |||
BC037091.1 | - | 7 | 848 | |||
BC040807.1 | - | 8 | 872 | |||
BC049948.1 | - | 8 | 789 | 3666660 | ||
BC056982.1 | - | 7 | 2268 | |||
CA983915.1 | - | 6 | 661 | |||
CX219422.1 | - | 3 | 333 |