Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plgrkt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026362.2 | NP_080638.1 | 5 | 494 | ||
GenBank mRNA | AK017171.1 | - | 5 | 519 | ||
AK017215.1 | - | 5 | 495 | |||
AK168454.1 | - | 5 | 470 | |||
AK170700.1 | - | 6 | 568 | |||
BC024953.1 | - | 5 | 453 | 4951101 |