Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026382.2 | NP_080658.2 | 8 | 854 | ||
GenBank mRNA | AK004216.1 | - | 2 | 184 | ||
AK007431.1 | - | 8 | 843 | |||
AK012916.1 | - | 8 | 849 | |||
AK018315.1 | - | 8 | 849 | |||
BC021443.1 | - | 7 | 648 | 5038605 | ||
BC037073.1 | - | 8 | 844 | |||
BY430458.1 | - | 2 | 73 |