Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026382.2 | NP_080658.2 | 5 | 515 | ||
GenBank mRNA | AK007431.1 | - | 5 | 504 | ||
AK012916.1 | - | 5 | 510 | |||
AK018315.1 | - | 5 | 510 | |||
AK167282.1 | - | 1 | 1105 | |||
BC037073.1 | - | 5 | 505 |