Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026390.2 | NP_080666.2 | 5 | 699 | ||
GenBank mRNA | AK011233.1 | - | 5 | 596 | ||
AK012824.1 | - | 5 | 593 | |||
AK019371.1 | - | 5 | 592 | |||
AK029431.1 | - | 5 | 745 | |||
AK032206.1 | - | 5 | 578 | |||
AK075716.1 | - | 5 | 434 | |||
AK145551.1 | - | 2 | 125 | |||
AK147795.1 | - | 5 | 572 | |||
AK148333.1 | - | 5 | 585 | |||
BC012271.1 | - | 5 | 711 | |||
BC019795.1 | - | 5 | 718 | 4158848 | ||
BC030190.1 | - | 5 | 708 |