Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175124.5 | NP_780333.3 | 6 | 546 | ||
GenBank mRNA | AK077270.1 | - | 6 | 542 | ||
AK159417.1 | - | 6 | 515 | |||
BC086694.1 | - | 6 | 437 | 30697866 |