Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175124.5 | NP_780333.3 | 8, 9 | 992 | ||
GenBank mRNA | AK004904.1 | - | 8, 9 | 865 | ||
AK006352.1 | - | 2, 3 | 198 | |||
AK077270.1 | - | 8, 9 | 988 | |||
AK159417.1 | - | 8, 9 | 961 | |||
BC027058.1 | - | 5, 6 | 549 | |||
BC052051.1 | - | 6, 7 | 763 | |||
BC086694.1 | - | 8, 9 | 883 | 30697866 |