Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285452.1 | NP_001272381.1 | 2 | 517 | ||
NM_028975.4 | NP_083251.2 | 3 | 360 | |||
NM_030108.2 | NP_084384.1 | 3 | 360 | |||
GenBank mRNA | AK005493.1 | - | 3 | 252 | ||
AK007607.1 | - | 3 | 168 | |||
AK007750.1 | - | 3 | 249 | |||
AK012413.1 | - | 3 | 274 | |||
AK017281.1 | - | 3 | 229 | |||
AK019127.1 | - | 2 | 186 | |||
AK032555.1 | - | 3 | 267 | |||
AK153441.1 | - | 3 | 185 | |||
AK165369.1 | - | 3 | 162 | |||
BC005562.1 | - | 3 | 161 | 3965864 | ||
BC016570.1 | - | 2 | 233 | 3583937 | ||
BC023966.1 | - | 2 | 226 | 5342147 | ||
CJ103460.1 | - | 2 | 278 |