Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198295.2 | NP_938037.2 | 14 | 1173 | ||
GenBank mRNA | AK035946.1 | - | 14 | 1121 | ||
AK042787.1 | - | 14 | 1116 | |||
AK048502.1 | - | 14 | 1129 | |||
AK129451.1 | - | 14 | 1067 | |||
AK140709.1 | - | 14 | 1174 | |||
AK157865.1 | - | 13 | 2230 | |||
BC057139.1 | - | 9 | 641 |