Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026512.1 | NP_080788.1 | 4 | 474 | ||
GenBank mRNA | AK002907.1 | - | 4 | 472 | ||
AK007606.1 | - | 4 | 468 | |||
AK008202.1 | - | 4 | 492 | |||
AK017801.1 | - | 4 | 477 | |||
AK078232.1 | - | 4 | 519 | |||
AK152712.1 | - | 4 | 484 | |||
AK170099.1 | - | 4 | 477 | |||
BC023146.1 | - | 4 | 459 | 3990118 | ||
BC025162.1 | - | 4 | 474 | 4921451 | ||
BC029322.1 | - | 2 | 109 |