Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026573.2 | NP_080849.1 | 1, 2 | 251 | ||
GenBank mRNA | AK011606.1 | - | 1, 2 | 249 | ||
AK019989.1 | - | 1, 2 | 229 | |||
AK145431.1 | - | 1, 2 | 254 | |||
AK162222.1 | - | 1, 2 | 247 | |||
BC058761.1 | - | 1, 2 | 232 | |||
BC117769.1 | - | 1, 2 | 269 | 40091025 | ||
BC117770.1 | - | 1, 2 | 269 | 40091027 |