Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026601.3 | NP_080877.3 | 6 | 740 | ||
GenBank mRNA | AK004328.1 | - | 5 | 328 | ||
AK020099.1 | - | 6 | 614 | |||
BC024619.1 | - | 6 | 728 | 4923664 | ||
BC100401.1 | - | 6 | 704 | |||
BX525112.1 | - | 4 | 181 | |||
CB952524.1 | - | 4 | 245 |